Assay Details
| Gene Symbol: ATP8A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105529.1 | NP_001098999.1 | 6 - 7 | 685 | 145 | |
| XM_005248043.2 | XP_005248100.1 | 6 - 7 | 680 | 145 | ||
| XM_011513615.1 | XP_011511917.1 | 6 - 7 | 680 | 145 | ||
| XM_011513618.1 | XP_011511920.1 | 6 - 7 | 680 | 145 | ||
| GenBank mRNA | AB209687.1 | - | 6 - 7 | 538 | 145 | |
| BC109317.1 | - | 6 - 7 | 447 | 40036536 | 145 | |
| BC109318.1 | - | 6 - 7 | 453 | 40036538 | 145 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001152.hg.1
Set Membership: |
|

ATP8A1
Genomic Map