Assay Details
| Gene Symbol: PKIG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001281444.1 | NP_001268373.1 | 2 - 3 | 345 | 135 | |
| NM_001281445.1 | NP_001268374.1 | 1 - 2 | 183 | 135 | ||
| NM_181804.2 | NP_861520.1 | 2 - 3 | 345 | 135 | ||
| GenBank mRNA | AA463287.1 | - | 1 - 2 | 183 | 135 | |
| AK309209.1 | - | 1 - 2 | 153 | 135 | ||
| BI461377.1 | - | 2 - 3 | 222 | 135 | ||
| BU161358.1 | - | 2 - 3 | 291 | 135 | ||
| CD244921.1 | - | 2 - 3 | 217 | 135 | ||
| DN994035.1 | - | 1 - 2 | 165 | 135 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_001281444.1,NM_001281445.1,NM_181804.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000349959.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000349959.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000349959.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000349959.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000349959.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372886.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372886.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372886.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372886.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372886.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372886.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372886.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372887.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372887.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372887.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372887.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372887.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372891.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372891.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372891.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372891.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372891.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372891.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372891.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372891.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372891.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372892.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372892.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372892.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372892.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372892.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372892.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372892.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372892.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372894.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372894.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372894.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372894.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372894.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372894.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000372894.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000477390.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000477390.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000477390.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | ENST00000477390.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_001281444.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_001281444.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_001281444.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_001281444.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_001281444.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_001281444.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_001281444.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_001281444.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_001281444.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_001281445.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_001281445.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_001281445.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_001281445.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_001281445.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_001281445.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_001281445.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_007066.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_007066.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_007066.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_007066.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_007066.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_007066.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_007066.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_181804.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_181804.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_181804.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_181804.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_181804.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_181804.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_181804.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_181804.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_181805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_181805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_181805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_181805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_181805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_181805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_181805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_181805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007457.hg.1 | NM_181805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000349959.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000349959.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000349959.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000349959.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000349959.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372886.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372886.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372886.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372886.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372886.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372886.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372886.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372887.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372887.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372887.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372887.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372887.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372891.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372891.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372891.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372891.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372891.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372891.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372891.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372891.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372891.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372892.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372892.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372892.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372892.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372892.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372892.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372892.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372892.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372894.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372894.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372894.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372894.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372894.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372894.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000372894.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000477390.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000477390.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000477390.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | ENST00000477390.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_001281444.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_001281444.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_001281444.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_001281444.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_001281444.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_001281444.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_001281444.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_001281444.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_001281444.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_001281445.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_001281445.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_001281445.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_001281445.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_001281445.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_001281445.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_001281445.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_007066.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_007066.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_007066.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_007066.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_007066.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_007066.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_007066.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_181804.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_181804.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_181804.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_181804.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_181804.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_181804.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_181804.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_181804.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_181805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_181805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_181805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_181805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_181805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_181805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_181805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_181805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007457.hg.1 | NM_181805.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000151758.hg.1 | ENST00000372886.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000151758.hg.1 | NM_001281445.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000151758.hg.1 | TC2000007457.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000151758.hg.1 | ENST00000372886.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000151758.hg.1 | NM_001281445.1 | Refseq |

PKIG
Genomic Map