Assay Details
| Gene Symbol: NRAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006175.4 | NP_006166.3 | 11 - 12 | 1296 | 159 | |
| XM_005269864.2 | XP_005269921.1 | 11 - 12 | 1272 | 159 | ||
| GenBank mRNA | AY081943.2 | - | 11 - 12 | 1273 | 159 | |
| AY177621.1 | - | 11 - 12 | 1249 | 159 | ||
More Information
Updated Mapping Information
Set Membership: |
|

NRAP
Genomic Map