Assay Details
| Gene Symbol: WASF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005267206.2 | XP_005267263.1 | 1 - 2 | 175 | 157 | |
| XM_017011444.1 | XP_016866933.1 | 1 - 2 | 175 | 157 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005267206.2, XM_017011444.1 transcripts.
Set Membership: |
|

WASF1
Genomic Map