Assay Details
| Gene Symbol: CRY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004075.4 | NP_004066.1 | 7 - 8 | 2026 | 129 | |
| XM_017018832.1 | XP_016874321.1 | 8 - 9 | 2161 | 129 | ||
| GenBank mRNA | AK098615.1 | - | 2 - 3 | 429 | 129 | |
| AK125915.1 | - | 9 - 10 | 1689 | 129 | ||
| AK290552.1 | - | 7 - 8 | 1326 | 129 | ||
| BC017466.1 | - | 7 - 8 | 1449 | 129 | ||
| BC030519.1 | - | 7 - 8 | 1699 | 5169081 | 129 | |
| D83702.1 | - | 7 - 8 | 1716 | 129 | ||
| D84657.1 | - | 7 - 8 | 1130 | 129 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12001925.hg
Additional Information:
This assay is in the 5'UTR region of the AK098615.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
gluconeogenesis
transcription, DNA-templated
DNA damage induced protein phosphorylation
blue light signaling pathway
protein-chromophore linkage
lipid storage
negative regulation of protein ubiquitination
response to insulin
circadian regulation of gene expression
response to glucagon
glucose homeostasis
regulation of circadian rhythm
negative regulation of circadian rhythm
entrainment of circadian clock by photoperiod
negative regulation of G-protein coupled receptor protein signaling pathway
negative regulation of transcription, DNA-templated
regulation of DNA damage checkpoint
negative regulation of glucocorticoid receptor signaling pathway
negative regulation of glucocorticoid secretion
nucleotide binding
transcription factor activity, transcription factor binding
core promoter sequence-specific DNA binding
core promoter binding
DNA binding
double-stranded DNA binding
deoxyribodipyrimidine photo-lyase activity
DNA (6-4) photolyase activity
protein binding
transcription factor binding
blue light photoreceptor activity
protein kinase binding
phosphatase binding
nuclear hormone receptor binding
histone deacetylase binding
ubiquitin binding
negative regulation of transcription from RNA polymerase II promoter
gluconeogenesis
transcription, DNA-templated
DNA damage induced protein phosphorylation
blue light signaling pathway
protein-chromophore linkage
lipid storage
negative regulation of protein ubiquitination
response to insulin
circadian regulation of gene expression
response to glucagon
glucose homeostasis
regulation of circadian rhythm
negative regulation of circadian rhythm
entrainment of circadian clock by photoperiod
negative regulation of G-protein coupled receptor protein signaling pathway
negative regulation of transcription, DNA-templated
regulation of DNA damage checkpoint
negative regulation of glucocorticoid receptor signaling pathway
negative regulation of glucocorticoid secretion
nucleotide binding
transcription factor activity, transcription factor binding
core promoter sequence-specific DNA binding
core promoter binding
DNA binding
double-stranded DNA binding
deoxyribodipyrimidine photo-lyase activity
DNA (6-4) photolyase activity
protein binding
transcription factor binding
blue light photoreceptor activity
protein kinase binding
phosphatase binding
nuclear hormone receptor binding
histone deacetylase binding
ubiquitin binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12025088.hg.1 | ENST00000008527.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12025088.hg.1 | ENST00000319645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12025088.hg.1 | NM_004075.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12025088.hg.1 | TC12001925.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12025091.hg.1 | ENST00000008527.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12025091.hg.1 | NM_004075.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12025091.hg.1 | TC12001925.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12025090.hg.1 | ENST00000008527.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12025090.hg.1 | ENST00000319645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12025090.hg.1 | NM_004075.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12025090.hg.1 | TC12001925.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | ENST00000008527.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | ENST00000008527.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | ENST00000008527.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | ENST00000008527.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | ENST00000008527.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | ENST00000008527.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | ENST00000008527.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | ENST00000008527.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | ENST00000008527.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | ENST00000008527.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | ENST00000008527.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | ENST00000008527.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | ENST00000008527.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | ENST00000008527.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | ENST00000008527.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | ENST00000008527.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | ENST00000008527.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | ENST00000008527.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | ENST00000546722.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | ENST00000546722.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | ENST00000546722.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | ENST00000546722.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | ENST00000546722.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | ENST00000549356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | ENST00000549356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | ENST00000549356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | ENST00000549356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | ENST00000549356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | ENST00000550633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | ENST00000550633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | ENST00000550633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | ENST00000552790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | ENST00000552790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | ENST00000552790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | ENST00000552790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | ENST00000552790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | ENST00000552790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | ENST00000552790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | ENST00000552790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | ENST00000552790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | ENST00000552790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | ENST00000552790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | ENST00000552790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | ENST00000552790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | ENST00000552790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | ENST00000552790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | ENST00000552790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | ENST00000552790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | ENST00000552790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | ENST00000552790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | ENST00000552790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | ENST00000552790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | NM_004075.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | NM_004075.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | NM_004075.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | NM_004075.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | NM_004075.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | NM_004075.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | NM_004075.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | NM_004075.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | NM_004075.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | NM_004075.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | NM_004075.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | NM_004075.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | NM_004075.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | NM_004075.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | NM_004075.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | NM_004075.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | NM_004075.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | NM_004075.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011812.hg.1 | NM_004075.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200191529.hg.1 | ENST00000008527.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191529.hg.1 | ENST00000552790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191529.hg.1 | NM_004075.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200191529.hg.1 | ENST00000008527.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191529.hg.1 | ENST00000552790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191529.hg.1 | NM_004075.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200191529.hg.1 | TC1200011812.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | ENST00000008527.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | ENST00000008527.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | ENST00000008527.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | ENST00000008527.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | ENST00000008527.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | ENST00000008527.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | ENST00000008527.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | ENST00000008527.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | ENST00000008527.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | ENST00000008527.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | ENST00000008527.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | ENST00000008527.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | ENST00000008527.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | ENST00000008527.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | ENST00000008527.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | ENST00000008527.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | ENST00000008527.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | ENST00000008527.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | ENST00000546722.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | ENST00000546722.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | ENST00000546722.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | ENST00000546722.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | ENST00000546722.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | ENST00000549356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | ENST00000549356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | ENST00000549356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | ENST00000549356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | ENST00000549356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | ENST00000550633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | ENST00000550633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | ENST00000550633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | ENST00000552790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | ENST00000552790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | ENST00000552790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | ENST00000552790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | ENST00000552790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | ENST00000552790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | ENST00000552790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | ENST00000552790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | ENST00000552790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | ENST00000552790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | ENST00000552790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | ENST00000552790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | ENST00000552790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | ENST00000552790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | ENST00000552790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | ENST00000552790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | ENST00000552790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | ENST00000552790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | ENST00000552790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | ENST00000552790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | ENST00000552790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | NM_004075.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | NM_004075.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | NM_004075.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | NM_004075.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | NM_004075.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | NM_004075.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | NM_004075.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | NM_004075.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | NM_004075.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | NM_004075.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | NM_004075.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | NM_004075.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | NM_004075.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | NM_004075.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | NM_004075.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | NM_004075.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | NM_004075.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | NM_004075.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011812.hg.1 | NM_004075.4 | Refseq |

CRY1
Genomic Map