Assay Details
| Gene Symbol: MXD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011513388.2 | XP_011511690.1 | 3 - 4 | 592 | 155 | |
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | ENST00000337190.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | ENST00000337190.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | ENST00000337190.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | ENST00000337190.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | ENST00000337190.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | ENST00000337190.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | ENST00000337190.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | ENST00000337190.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | ENST00000337190.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | ENST00000337190.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | ENST00000337190.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | ENST00000337190.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | ENST00000337190.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | ENST00000337190.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | ENST00000337190.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | ENST00000337190.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | ENST00000510822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | ENST00000510822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | ENST00000510822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | ENST00000510822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | ENST00000510822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | ENST00000510822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | ENST00000510822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | ENST00000510822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | ENST00000513372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | ENST00000513372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | ENST00000513372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | ENST00000513372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | ENST00000513372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | ENST00000513372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | ENST00000513372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | ENST00000513372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | ENST00000513372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | ENST00000513372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | ENST00000513372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | ENST00000513380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | ENST00000513380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | ENST00000513380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | ENST00000513380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | ENST00000513380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | ENST00000513380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | ENST00000513380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | ENST00000513380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | ENST00000515378.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | ENST00000515378.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | ENST00000515378.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | ENST00000515378.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | ENST00000515378.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | ENST00000515378.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | ENST00000515378.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | ENST00000515378.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | ENST00000537353.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | NM_006454.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | NM_006454.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | NM_006454.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | NM_006454.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | NM_006454.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | NM_006454.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | NM_006454.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | NM_006454.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | NM_006454.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | NM_006454.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | NM_006454.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | NM_006454.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | NM_006454.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | NM_006454.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009765.hg.1 | NR_039964.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | ENST00000337190.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | ENST00000337190.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | ENST00000337190.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | ENST00000337190.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | ENST00000337190.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | ENST00000337190.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | ENST00000337190.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | ENST00000337190.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | ENST00000337190.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | ENST00000337190.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | ENST00000337190.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | ENST00000337190.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | ENST00000337190.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | ENST00000337190.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | ENST00000337190.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | ENST00000337190.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | ENST00000510822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | ENST00000510822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | ENST00000510822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | ENST00000510822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | ENST00000510822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | ENST00000510822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | ENST00000510822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | ENST00000510822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | ENST00000513372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | ENST00000513372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | ENST00000513372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | ENST00000513372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | ENST00000513372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | ENST00000513372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | ENST00000513372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | ENST00000513372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | ENST00000513372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | ENST00000513372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | ENST00000513372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | ENST00000513380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | ENST00000513380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | ENST00000513380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | ENST00000513380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | ENST00000513380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | ENST00000513380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | ENST00000513380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | ENST00000513380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | ENST00000515378.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | ENST00000515378.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | ENST00000515378.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | ENST00000515378.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | ENST00000515378.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | ENST00000515378.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | ENST00000515378.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | ENST00000515378.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | ENST00000537353.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | NM_006454.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | NM_006454.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | NM_006454.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | NM_006454.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | NM_006454.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | NM_006454.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | NM_006454.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | NM_006454.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | NM_006454.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | NM_006454.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | NM_006454.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | NM_006454.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | NM_006454.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | NM_006454.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009765.hg.1 | NR_039964.1 | Refseq |

MXD4
Genomic Map