| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012431.hg.1 |
ENST00000518143.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012431.hg.1 |
ENST00000518143.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012431.hg.1 |
ENST00000518143.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012431.hg.1 |
ENST00000518143.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012431.hg.1 |
ENST00000518143.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012431.hg.1 |
ENST00000518143.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012431.hg.1 |
ENST00000518143.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012431.hg.1 |
ENST00000518143.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012431.hg.1 |
ENST00000518143.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012431.hg.1 |
ENST00000521894.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012431.hg.1 |
ENST00000521894.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012431.hg.1 |
ENST00000521894.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012431.hg.1 |
ENST00000521894.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012431.hg.1 |
ENST00000521894.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012431.hg.1 |
ENST00000522357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012431.hg.1 |
ENST00000522357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012431.hg.1 |
ENST00000522357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012431.hg.1 |
ENST00000523977.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012431.hg.1 |
ENST00000523977.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012431.hg.1 |
NR_024360.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012431.hg.1 |
NR_024360.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012431.hg.1 |
NR_024360.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012431.hg.1 |
NR_024360.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012431.hg.1 |
NR_024360.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012431.hg.1 |
NR_024360.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012431.hg.1 |
NR_024360.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012431.hg.1 |
NR_024360.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012431.hg.1 |
NR_024360.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08017317.hg.1 |
TC08001345.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08017317.hg.1 |
ENST00000518143.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08017317.hg.1 |
NR_024360.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08017315.hg.1 |
TC08001345.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08017315.hg.1 |
ENST00000518143.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08017315.hg.1 |
NR_024360.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08017316.hg.1 |
TC08001345.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08017316.hg.1 |
ENST00000518143.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08017316.hg.1 |
ENST00000522961.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08017316.hg.1 |
NR_024360.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800187435.hg.1 |
ENST00000518143.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800187435.hg.1 |
NR_024360.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800187435.hg.1 |
ENST00000518143.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800187435.hg.1 |
NR_024360.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800187435.hg.1 |
TC0800012431.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0800012431.hg.1 |
ENST00000518143.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012431.hg.1 |
ENST00000518143.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012431.hg.1 |
ENST00000518143.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012431.hg.1 |
ENST00000518143.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012431.hg.1 |
ENST00000518143.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012431.hg.1 |
ENST00000518143.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012431.hg.1 |
ENST00000518143.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012431.hg.1 |
ENST00000518143.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012431.hg.1 |
ENST00000518143.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012431.hg.1 |
ENST00000521894.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012431.hg.1 |
ENST00000521894.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012431.hg.1 |
ENST00000521894.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012431.hg.1 |
ENST00000521894.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012431.hg.1 |
ENST00000521894.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012431.hg.1 |
ENST00000522357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012431.hg.1 |
ENST00000522357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012431.hg.1 |
ENST00000522357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012431.hg.1 |
ENST00000523977.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012431.hg.1 |
ENST00000523977.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012431.hg.1 |
NR_024360.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012431.hg.1 |
NR_024360.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012431.hg.1 |
NR_024360.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012431.hg.1 |
NR_024360.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012431.hg.1 |
NR_024360.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012431.hg.1 |
NR_024360.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012431.hg.1 |
NR_024360.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012431.hg.1 |
NR_024360.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012431.hg.1 |
NR_024360.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800187437.hg.1 |
ENST00000518143.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800187437.hg.1 |
NR_024360.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800187437.hg.1 |
ENST00000518143.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800187437.hg.1 |
NR_024360.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800187437.hg.1 |
TC0800012431.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0800187436.hg.1 |
ENST00000518143.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800187436.hg.1 |
NR_024360.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800187436.hg.1 |
ENST00000518143.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800187436.hg.1 |
NR_024360.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800187436.hg.1 |
TC0800012431.hg.1 |
Clariom D TC |