Assay Details
| Gene Symbol: TSPYL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022117.3 | NP_071400.1 | 3 - 4 | 1137 | 163 | |
| XM_006724592.3 | XP_006724655.1 | 3 - 4 | 1170 | 163 | ||
| XM_017029726.1 | XP_016885215.1 | 2 - 3 | 1302 | 163 | ||
| XM_017029727.1 | XP_016885216.1 | 2 - 3 | 1302 | 163 | ||
| GenBank mRNA | AB015345.1 | - | 3 - 4 | 377 | 163 | |
| AF254794.1 | - | 3 - 4 | 876 | 163 | ||
| AF273046.1 | - | 3 - 4 | 1127 | 163 | ||
| AK131220.1 | - | 2 - 3 | 1275 | 163 | ||
| AK292462.1 | - | 3 - 4 | 1130 | 163 | ||
| AL713652.1 | - | 3 - 4 | 1116 | 163 | ||
| AY040871.1 | - | 3 - 4 | 1126 | 163 | ||
| BC024270.1 | - | 3 - 4 | 1117 | 2819684 | 163 | |
More Information
Set Membership: |
|

TSPYL2
Genomic Map