Assay Details
| Gene Symbol: NT5C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011524700.2 | XP_011523002.2 | 1 | 264 | 159 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | armadillo repeat containing 7 | ARMC7 | -1.3kb | Anti-sense | Chr. 17 - 75109952 - 75130265 on NCBI Build GRCh38 | |
| Downstream | hematological and neurological expressed 1 | HN1 |
ARM2, HN1A |
3.3kb | Sense | Chr. 17 - 75135243 - 75154683 on NCBI Build GRCh38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011524700.2 transcripts.
Set Membership: |
|

NT5C
Genomic Map