| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010421.hg.1 |
ENST00000379811.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010421.hg.1 |
ENST00000379811.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010421.hg.1 |
ENST00000379811.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010421.hg.1 |
ENST00000379811.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010421.hg.1 |
ENST00000379811.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010421.hg.1 |
ENST00000379811.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010421.hg.1 |
ENST00000379811.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010421.hg.1 |
ENST00000444837.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010421.hg.1 |
ENST00000444837.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010421.hg.1 |
ENST00000444837.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010421.hg.1 |
ENST00000444837.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010421.hg.1 |
ENST00000444837.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010421.hg.1 |
ENST00000444837.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010421.hg.1 |
ENST00000444837.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010421.hg.1 |
ENST00000568675.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010421.hg.1 |
ENST00000568675.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010421.hg.1 |
ENST00000568675.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010421.hg.1 |
ENST00000568675.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010421.hg.1 |
ENST00000568675.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010421.hg.1 |
ENST00000568675.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010421.hg.1 |
ENST00000569500.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010421.hg.1 |
ENST00000569500.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010421.hg.1 |
ENST00000569500.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010421.hg.1 |
ENST00000569500.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010421.hg.1 |
ENST00000569500.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010421.hg.1 |
NM_001301267.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010421.hg.1 |
NM_001301267.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010421.hg.1 |
NM_001301267.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010421.hg.1 |
NM_001301267.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010421.hg.1 |
NM_001301267.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010421.hg.1 |
NM_001301267.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010421.hg.1 |
NM_001301267.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010421.hg.1 |
NM_005950.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010421.hg.1 |
NM_005950.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010421.hg.1 |
NM_005950.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010421.hg.1 |
NM_005950.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010421.hg.1 |
NM_005950.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010421.hg.1 |
NM_005950.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010421.hg.1 |
NM_005950.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010421.hg.1 |
ENST00000379811.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010421.hg.1 |
ENST00000379811.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010421.hg.1 |
ENST00000379811.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010421.hg.1 |
ENST00000379811.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010421.hg.1 |
ENST00000379811.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010421.hg.1 |
ENST00000379811.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010421.hg.1 |
ENST00000379811.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010421.hg.1 |
ENST00000444837.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010421.hg.1 |
ENST00000444837.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010421.hg.1 |
ENST00000444837.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010421.hg.1 |
ENST00000444837.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010421.hg.1 |
ENST00000444837.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010421.hg.1 |
ENST00000444837.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010421.hg.1 |
ENST00000444837.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010421.hg.1 |
ENST00000568675.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010421.hg.1 |
ENST00000568675.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010421.hg.1 |
ENST00000568675.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010421.hg.1 |
ENST00000568675.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010421.hg.1 |
ENST00000568675.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010421.hg.1 |
ENST00000568675.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010421.hg.1 |
ENST00000569500.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010421.hg.1 |
ENST00000569500.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010421.hg.1 |
ENST00000569500.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010421.hg.1 |
ENST00000569500.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010421.hg.1 |
ENST00000569500.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010421.hg.1 |
NM_001301267.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010421.hg.1 |
NM_001301267.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010421.hg.1 |
NM_001301267.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010421.hg.1 |
NM_001301267.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010421.hg.1 |
NM_001301267.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010421.hg.1 |
NM_001301267.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010421.hg.1 |
NM_001301267.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010421.hg.1 |
NM_005950.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010421.hg.1 |
NM_005950.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010421.hg.1 |
NM_005950.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010421.hg.1 |
NM_005950.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010421.hg.1 |
NM_005950.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010421.hg.1 |
NM_005950.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010421.hg.1 |
NM_005950.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600180008.hg.1 |
ENST00000379811.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600180008.hg.1 |
ENST00000444837.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600180008.hg.1 |
ENST00000569500.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600180008.hg.1 |
NM_001301267.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600180008.hg.1 |
NM_005950.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600180008.hg.1 |
ENST00000379811.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600180008.hg.1 |
ENST00000444837.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600180008.hg.1 |
ENST00000569500.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600180008.hg.1 |
NM_001301267.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600180008.hg.1 |
NM_005950.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600180008.hg.1 |
TC1600010421.hg.1 |
Clariom D TC |