Assay Details
| Gene Symbol: DIRAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145173.3 | NP_660156.1 | 2 | 2183 | 136 | |
| XM_011527713.2 | XP_011526015.1 | 2 | 2348 | 136 | ||
| GenBank mRNA | AK124445.1 | - | 1 | 1494 | 136 | |
| AL833958.1 | - | 1 | 972 | 136 | ||
| BC030660.1 | - | 2 | 2162 | 4814337 | 136 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

DIRAS1
Genomic Map