Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X014608.hg.1 |
TC0X001072.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X014608.hg.1 |
ENST00000328235.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X014608.hg.1 |
ENST00000375180.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X014608.hg.1 |
NM_017848.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00166955.hg.1 |
ENST00000328235.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00166955.hg.1 |
ENST00000375180.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00166955.hg.1 |
NM_001300788.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00166955.hg.1 |
NM_017848.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00166955.hg.1 |
ENST00000328235.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00166955.hg.1 |
ENST00000375180.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00166955.hg.1 |
NM_001300788.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00166955.hg.1 |
NM_017848.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00166955.hg.1 |
TC0X00009776.hg.1 |
Clariom D TC |