Assay Details
| Gene Symbol: FAM120C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300788.1 | NP_001287717.1 | 6 - 7 | 1425 | 75 | |
| NM_017848.5 | NP_060318.4 | 6 - 7 | 1402 | 75 | ||
| XM_006724589.3 | XP_006724652.1 | 6 - 7 | 1398 | 75 | ||
| GenBank mRNA | AY150025.1 | - | 6 - 7 | 1402 | 75 | |
| BC136413.1 | - | 6 - 7 | 1399 | 9020401 | 75 | |
| BC144081.1 | - | 6 - 7 | 1399 | 75 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X001072.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X014615.hg.1 | TC0X001072.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X014615.hg.1 | ENST00000328235.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X014615.hg.1 | ENST00000375180.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X014615.hg.1 | NM_017848.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | ENST00000328235.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | ENST00000328235.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | ENST00000328235.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | ENST00000328235.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | ENST00000328235.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | ENST00000328235.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | ENST00000328235.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | ENST00000328235.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | ENST00000328235.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | ENST00000328235.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | ENST00000328235.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | ENST00000328235.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | ENST00000328235.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | ENST00000328235.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | ENST00000328235.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | ENST00000328235.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | ENST00000328235.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | ENST00000328235.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | ENST00000375180.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | ENST00000375180.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | ENST00000375180.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | ENST00000375180.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | ENST00000375180.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | ENST00000375180.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | ENST00000375180.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | ENST00000375180.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | ENST00000375180.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | ENST00000375180.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | ENST00000375180.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | ENST00000375180.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | ENST00000375180.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | ENST00000375180.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | ENST00000375180.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | ENST00000375180.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | ENST00000375180.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | ENST00000375180.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | ENST00000375180.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | ENST00000375180.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | ENST00000375180.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | ENST00000375180.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | ENST00000375180.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | ENST00000477084.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | ENST00000477084.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | ENST00000477084.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | ENST00000477084.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | ENST00000497680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | ENST00000497680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | NM_001300788.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | NM_001300788.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | NM_001300788.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | NM_001300788.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | NM_001300788.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | NM_001300788.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | NM_001300788.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | NM_001300788.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | NM_001300788.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | NM_001300788.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | NM_001300788.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | NM_001300788.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | NM_001300788.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | NM_001300788.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | NM_001300788.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | NM_001300788.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | NM_001300788.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | NM_001300788.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | NM_001300788.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | NM_001300788.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | NM_001300788.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | NM_017848.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | NM_017848.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | NM_017848.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | NM_017848.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | NM_017848.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | NM_017848.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | NM_017848.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | NM_017848.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | NM_017848.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | NM_017848.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | NM_017848.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | NM_017848.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | NM_017848.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | NM_017848.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | NM_017848.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | NM_017848.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | NM_017848.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | NM_017848.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | NM_017848.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | NM_017848.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | NM_017848.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | NM_017848.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | NM_198456.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | NM_198456.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | NM_198456.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009776.hg.1 | NM_198456.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00166962.hg.1 | ENST00000328235.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00166962.hg.1 | ENST00000375180.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00166962.hg.1 | NM_001300788.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00166962.hg.1 | NM_017848.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00166962.hg.1 | ENST00000328235.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00166962.hg.1 | ENST00000375180.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00166962.hg.1 | NM_001300788.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00166962.hg.1 | NM_017848.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00166962.hg.1 | TC0X00009776.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | ENST00000328235.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | ENST00000328235.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | ENST00000328235.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | ENST00000328235.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | ENST00000328235.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | ENST00000328235.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | ENST00000328235.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | ENST00000328235.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | ENST00000328235.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | ENST00000328235.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | ENST00000328235.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | ENST00000328235.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | ENST00000328235.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | ENST00000328235.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | ENST00000328235.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | ENST00000328235.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | ENST00000328235.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | ENST00000328235.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | ENST00000375180.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | ENST00000375180.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | ENST00000375180.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | ENST00000375180.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | ENST00000375180.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | ENST00000375180.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | ENST00000375180.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | ENST00000375180.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | ENST00000375180.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | ENST00000375180.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | ENST00000375180.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | ENST00000375180.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | ENST00000375180.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | ENST00000375180.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | ENST00000375180.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | ENST00000375180.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | ENST00000375180.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | ENST00000375180.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | ENST00000375180.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | ENST00000375180.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | ENST00000375180.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | ENST00000375180.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | ENST00000375180.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | ENST00000477084.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | ENST00000477084.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | ENST00000477084.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | ENST00000477084.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | ENST00000497680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | ENST00000497680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | NM_001300788.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | NM_001300788.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | NM_001300788.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | NM_001300788.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | NM_001300788.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | NM_001300788.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | NM_001300788.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | NM_001300788.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | NM_001300788.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | NM_001300788.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | NM_001300788.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | NM_001300788.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | NM_001300788.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | NM_001300788.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | NM_001300788.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | NM_001300788.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | NM_001300788.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | NM_001300788.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | NM_001300788.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | NM_001300788.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | NM_001300788.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | NM_017848.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | NM_017848.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | NM_017848.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | NM_017848.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | NM_017848.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | NM_017848.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | NM_017848.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | NM_017848.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | NM_017848.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | NM_017848.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | NM_017848.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | NM_017848.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | NM_017848.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | NM_017848.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | NM_017848.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | NM_017848.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | NM_017848.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | NM_017848.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | NM_017848.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | NM_017848.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | NM_017848.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | NM_017848.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | NM_198456.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | NM_198456.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | NM_198456.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009776.hg.1 | NM_198456.2 | Refseq |

FAM120C
Genomic Map