Assay Details
Gene Symbol: KRTAP10-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181688.2 | NP_859016.1 | 1 | 946 | 159 | |
GenBank mRNA | AJ566387.1 | - | 1 | 946 | 159 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
