Assay Details
| Gene Symbol: ENTPD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198585.2 | NP_940987.2 | 7 - 8 | 1233 | 113 | |
| XM_011518666.2 | XP_011516968.1 | 8 - 9 | 1549 | 113 | ||
| GenBank mRNA | AY359088.1 | - | 7 - 8 | 1233 | 113 | |
| BC141810.1 | - | 7 - 8 | 1134 | 40147640 | 113 | |
More Information
Set Membership: |
|

ENTPD8
Genomic Map