Assay Details
Gene Symbol: AQP12A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198998.2 | NP_945349.1 | 1 - 2 | 197 | 87 | |
GenBank mRNA | AB040748.1 | - | 1 - 2 | 197 | 87 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02001470.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR02023654.hg.1 | TC02001470.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02023654.hg.1 | ENST00000337801.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02023654.hg.1 | ENST00000429564.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02023654.hg.1 | NM_198998.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02023659.hg.1 | TC02001470.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02023659.hg.1 | ENST00000337801.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02023659.hg.1 | ENST00000373309.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02023659.hg.1 | ENST00000429564.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02023659.hg.1 | NM_198998.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | ENST00000337801.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | ENST00000337801.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | ENST00000337801.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | ENST00000337801.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | ENST00000337801.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | ENST00000337801.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | ENST00000429564.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | ENST00000429564.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | ENST00000429564.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | ENST00000429564.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | ENST00000429564.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | ENST00000429564.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | ENST00000429564.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | ENST00000460527.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | ENST00000460527.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | ENST00000460527.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | ENST00000460527.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | ENST00000460527.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | ENST00000471878.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | ENST00000471878.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | ENST00000471878.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | ENST00000471878.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | ENST00000474778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | ENST00000474778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | ENST00000474778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | NM_198998.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | NM_198998.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | NM_198998.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | NM_198998.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | NM_198998.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011372.hg.1 | NM_198998.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02023651.hg.1 | TC02001470.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02023651.hg.1 | ENST00000337801.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02023651.hg.1 | ENST00000373309.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02023651.hg.1 | ENST00000429564.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02023651.hg.1 | NM_198998.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02023653.hg.1 | TC02001470.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02023653.hg.1 | ENST00000337801.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02023653.hg.1 | ENST00000429564.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02023653.hg.1 | NM_198998.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02023658.hg.1 | TC02001470.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02023658.hg.1 | ENST00000337801.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02023658.hg.1 | ENST00000429564.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02023658.hg.1 | NM_198998.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200182065.hg.1 | ENST00000337801.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200182065.hg.1 | ENST00000429564.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200182065.hg.1 | NM_198998.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200182065.hg.1 | ENST00000337801.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200182065.hg.1 | ENST00000429564.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200182065.hg.1 | NM_198998.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200182065.hg.1 | TC0200011372.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | ENST00000337801.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | ENST00000337801.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | ENST00000337801.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | ENST00000337801.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | ENST00000337801.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | ENST00000337801.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | ENST00000429564.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | ENST00000429564.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | ENST00000429564.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | ENST00000429564.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | ENST00000429564.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | ENST00000429564.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | ENST00000429564.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | ENST00000460527.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | ENST00000460527.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | ENST00000460527.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | ENST00000460527.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | ENST00000460527.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | ENST00000471878.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | ENST00000471878.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | ENST00000471878.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | ENST00000471878.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | ENST00000474778.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | ENST00000474778.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | ENST00000474778.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | NM_198998.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | NM_198998.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | NM_198998.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | NM_198998.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | NM_198998.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011372.hg.1 | NM_198998.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200182064.hg.1 | ENST00000337801.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200182064.hg.1 | ENST00000429564.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200182064.hg.1 | NM_198998.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200182064.hg.1 | ENST00000337801.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200182064.hg.1 | ENST00000429564.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200182064.hg.1 | NM_198998.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200182064.hg.1 | TC0200011372.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200182067.hg.1 | ENST00000337801.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200182067.hg.1 | ENST00000429564.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200182067.hg.1 | NM_198998.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200182067.hg.1 | ENST00000337801.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200182067.hg.1 | ENST00000429564.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200182067.hg.1 | NM_198998.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200182067.hg.1 | TC0200011372.hg.1 | Clariom D TC |
