| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008906.hg.1 |
ENST00000599172.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008906.hg.1 |
ENST00000599172.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008906.hg.1 |
ENST00000599172.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008906.hg.1 |
ENST00000599172.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011676.hg.1 |
ENST00000372491.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011676.hg.1 |
ENST00000372491.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011676.hg.1 |
ENST00000372491.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011676.hg.1 |
ENST00000372491.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011676.hg.1 |
ENST00000372491.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011676.hg.1 |
ENST00000372491.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011676.hg.1 |
NM_203434.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011676.hg.1 |
NM_203434.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011676.hg.1 |
NM_203434.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011676.hg.1 |
NM_203434.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011676.hg.1 |
NM_203434.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011676.hg.1 |
NM_203434.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900179931.hg.1 |
ENST00000372491.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900179931.hg.1 |
NM_203434.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900179931.hg.1 |
ENST00000372491.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900179931.hg.1 |
NM_203434.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900179931.hg.1 |
TC0900011676.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0900011676.hg.1 |
ENST00000372491.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011676.hg.1 |
ENST00000372491.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011676.hg.1 |
ENST00000372491.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011676.hg.1 |
ENST00000372491.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011676.hg.1 |
ENST00000372491.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011676.hg.1 |
ENST00000372491.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011676.hg.1 |
NM_203434.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011676.hg.1 |
NM_203434.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011676.hg.1 |
NM_203434.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011676.hg.1 |
NM_203434.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011676.hg.1 |
NM_203434.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011676.hg.1 |
NM_203434.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900179934.hg.1 |
ENST00000372491.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900179934.hg.1 |
NM_203434.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900179934.hg.1 |
ENST00000372491.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900179934.hg.1 |
NM_203434.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900179934.hg.1 |
TC0900011676.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0900008906.hg.1 |
ENST00000599172.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008906.hg.1 |
ENST00000599172.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008906.hg.1 |
ENST00000599172.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008906.hg.1 |
ENST00000599172.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900179933.hg.1 |
ENST00000372491.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900179933.hg.1 |
NM_203434.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900179933.hg.1 |
ENST00000372491.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900179933.hg.1 |
NM_203434.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900179933.hg.1 |
TC0900011676.hg.1 |
Clariom D TC |