Assay Details
Gene Symbol: ACSM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207321.2 | NP_997204.2 | 1 - 2 | 109 | 85 | |
GenBank mRNA | AK126134.1 | - | 1 - 2 | 109 | 85 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10000676.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK126134.1,NM_207321.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR10007748.hg.1 | ENST00000341686.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10007748.hg.1 | ENST00000430183.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10007748.hg.1 | ENST00000539707.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10007748.hg.1 | NM_207321.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10007748.hg.1 | TC10000676.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | ENST00000327739.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | ENST00000327739.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | ENST00000327739.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | ENST00000327739.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | ENST00000327739.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | ENST00000327739.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | ENST00000327739.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | ENST00000327739.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | ENST00000327739.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | ENST00000327739.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | ENST00000327739.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | ENST00000341686.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | ENST00000341686.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | ENST00000341686.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | ENST00000341686.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | ENST00000341686.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | ENST00000341686.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | ENST00000341686.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | ENST00000341686.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | ENST00000341686.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | ENST00000341686.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | ENST00000341686.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | ENST00000341686.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | ENST00000341686.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | ENST00000394005.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | ENST00000394005.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | ENST00000394005.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | ENST00000394005.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | ENST00000394005.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | ENST00000394005.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | ENST00000394005.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | ENST00000394005.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | ENST00000394005.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | ENST00000394005.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | ENST00000394005.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | ENST00000394005.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | ENST00000404473.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | ENST00000404473.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | ENST00000404473.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | ENST00000404473.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | ENST00000404473.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | ENST00000404473.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | ENST00000404473.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | ENST00000404473.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | ENST00000404473.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | ENST00000404473.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | NM_207321.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | NM_207321.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | NM_207321.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | NM_207321.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | NM_207321.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | NM_207321.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | NM_207321.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | NM_207321.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | NM_207321.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | NM_207321.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | NM_207321.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | NM_207321.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008516.hg.1 | NM_207321.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10007750.hg.1 | ENST00000341686.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10007750.hg.1 | ENST00000394005.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10007750.hg.1 | ENST00000430183.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10007750.hg.1 | ENST00000539707.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10007750.hg.1 | NM_207321.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10007750.hg.1 | TC10000676.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR10007754.hg.1 | ENST00000341686.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10007754.hg.1 | ENST00000394005.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10007754.hg.1 | ENST00000430183.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10007754.hg.1 | ENST00000539707.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10007754.hg.1 | NM_207321.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10007754.hg.1 | TC10000676.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1000158020.hg.1 | ENST00000327739.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000158020.hg.1 | ENST00000341686.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000158020.hg.1 | NM_207321.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000158020.hg.1 | ENST00000327739.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000158020.hg.1 | ENST00000341686.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000158020.hg.1 | NM_207321.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000158020.hg.1 | TC1000008516.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | ENST00000327739.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | ENST00000327739.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | ENST00000327739.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | ENST00000327739.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | ENST00000327739.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | ENST00000327739.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | ENST00000327739.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | ENST00000327739.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | ENST00000327739.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | ENST00000327739.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | ENST00000327739.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | ENST00000341686.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | ENST00000341686.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | ENST00000341686.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | ENST00000341686.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | ENST00000341686.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | ENST00000341686.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | ENST00000341686.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | ENST00000341686.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | ENST00000341686.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | ENST00000341686.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | ENST00000341686.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | ENST00000341686.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | ENST00000341686.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | ENST00000394005.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | ENST00000394005.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | ENST00000394005.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | ENST00000394005.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | ENST00000394005.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | ENST00000394005.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | ENST00000394005.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | ENST00000394005.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | ENST00000394005.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | ENST00000394005.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | ENST00000394005.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | ENST00000394005.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | ENST00000404473.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | ENST00000404473.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | ENST00000404473.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | ENST00000404473.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | ENST00000404473.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | ENST00000404473.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | ENST00000404473.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | ENST00000404473.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | ENST00000404473.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | ENST00000404473.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | NM_207321.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | NM_207321.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | NM_207321.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | NM_207321.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | NM_207321.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | NM_207321.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | NM_207321.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | NM_207321.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | NM_207321.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | NM_207321.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | NM_207321.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | NM_207321.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008516.hg.1 | NM_207321.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000158023.hg.1 | ENST00000327739.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000158023.hg.1 | ENST00000341686.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000158023.hg.1 | ENST00000394005.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000158023.hg.1 | ENST00000404473.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000158023.hg.1 | NM_207321.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000158023.hg.1 | ENST00000327739.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000158023.hg.1 | ENST00000341686.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000158023.hg.1 | ENST00000394005.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000158023.hg.1 | ENST00000404473.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000158023.hg.1 | NM_207321.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000158023.hg.1 | TC1000008516.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000158025.hg.1 | ENST00000327739.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000158025.hg.1 | ENST00000341686.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000158025.hg.1 | ENST00000394005.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000158025.hg.1 | ENST00000404473.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000158025.hg.1 | NM_207321.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000158025.hg.1 | ENST00000327739.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000158025.hg.1 | ENST00000341686.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000158025.hg.1 | ENST00000394005.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000158025.hg.1 | ENST00000404473.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000158025.hg.1 | NM_207321.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000158025.hg.1 | TC1000008516.hg.1 | Clariom D TC |
