Assay Details
Gene Symbol: CYP4V2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207352.3 | NP_997235.3 | 1 - 2 | 510 | 182 | |
XM_005262935.3 | XP_005262992.1 | 1 - 2 | 295 | 182 | ||
XM_017008037.1 | XP_016863526.1 | 1 - 2 | 354 | 182 | ||
GenBank mRNA | AK122600.1 | - | 1 - 2 | 245 | 182 | |
AK126473.1 | - | 2 - 3 | 179 | 116 | ||
AY422002.1 | - | 1 - 2 | 510 | 182 | ||
BC060857.1 | - | 1 - 2 | 376 | 30333559 | 182 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC04002914.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_017008037.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR04012492.hg.1 | TC04002914.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04012492.hg.1 | ENST00000274118.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04012492.hg.1 | ENST00000378802.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04012492.hg.1 | NM_207352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04012488.hg.1 | TC04002914.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04012488.hg.1 | ENST00000378802.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04012488.hg.1 | NM_207352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04012490.hg.1 | TC04002914.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04012490.hg.1 | ENST00000274118.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04012490.hg.1 | ENST00000378802.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04012490.hg.1 | NM_207352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04012487.hg.1 | TC04002914.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04012487.hg.1 | ENST00000378802.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04012487.hg.1 | NM_207352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04012489.hg.1 | TC04002914.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04012489.hg.1 | ENST00000378802.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04012489.hg.1 | NM_207352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000513354.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000513354.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000513354.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000513354.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000513354.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000513354.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000513354.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | NM_207352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | NM_207352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | NM_207352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | NM_207352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | NM_207352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | NM_207352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | NM_207352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | NM_207352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | NM_207352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | NM_207352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | NM_207352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | NM_207352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | NM_207352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | NM_207352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | NM_207352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | NM_207352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | NM_207352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04012494.hg.1 | TC04002914.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04012494.hg.1 | ENST00000274118.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04012494.hg.1 | ENST00000378802.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04012494.hg.1 | NM_207352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04012491.hg.1 | TC04002914.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04012491.hg.1 | ENST00000378802.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04012491.hg.1 | NM_207352.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400189388.hg.1 | ENST00000378802.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400189388.hg.1 | NM_207352.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400189388.hg.1 | ENST00000378802.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400189388.hg.1 | NM_207352.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400189388.hg.1 | TC0400012874.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000513354.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000513354.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000513354.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000513354.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000513354.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000513354.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000513354.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | NM_207352.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | NM_207352.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | NM_207352.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | NM_207352.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | NM_207352.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | NM_207352.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | NM_207352.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | NM_207352.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | NM_207352.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | NM_207352.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | NM_207352.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | NM_207352.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | NM_207352.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | NM_207352.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | NM_207352.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | NM_207352.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | NM_207352.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400189389.hg.1 | ENST00000378802.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400189389.hg.1 | NM_207352.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400189389.hg.1 | ENST00000378802.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400189389.hg.1 | NM_207352.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400189389.hg.1 | TC0400012874.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400189386.hg.1 | ENST00000378802.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400189386.hg.1 | NM_207352.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400189386.hg.1 | ENST00000378802.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400189386.hg.1 | NM_207352.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400189386.hg.1 | TC0400012874.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400189387.hg.1 | ENST00000378802.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400189387.hg.1 | NM_207352.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400189387.hg.1 | ENST00000378802.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400189387.hg.1 | NM_207352.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400189387.hg.1 | TC0400012874.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 104577 | - | - |
