Assay Details
| Gene Symbol: IFITM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003641.3 | NP_003632.3 | 1 - 2 | 368 | 180 | |
| GenBank mRNA | AK290480.1 | - | 1 - 2 | 343 | 180 | |
| BC000897.1 | - | 1 - 2 | 318 | 3464598 | 180 | |
| BT007173.1 | - | 1 - 2 | 188 | 180 | ||
| J04164.1 | - | 1 - 2 | 504 | 180 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11000011.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
ossification
cell surface receptor signaling pathway
negative regulation of cell proliferation
response to virus
negative regulation of cell migration
response to interferon-gamma
response to interferon-alpha
response to interferon-beta
negative regulation of viral genome replication
positive regulation of osteoblast differentiation
negative regulation of viral entry into host cell
regulation of immune response
defense response to virus
type I interferon signaling pathway
receptor signaling protein activity
protein binding
ossification
cell surface receptor signaling pathway
negative regulation of cell proliferation
response to virus
negative regulation of cell migration
response to interferon-gamma
response to interferon-alpha
response to interferon-beta
negative regulation of viral genome replication
positive regulation of osteoblast differentiation
negative regulation of viral entry into host cell
regulation of immune response
defense response to virus
type I interferon signaling pathway
receptor signaling protein activity
protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11000171.hg.1 | ENST00000328221.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11000171.hg.1 | ENST00000408968.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11000171.hg.1 | ENST00000452428.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11000171.hg.1 | ENST00000525554.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11000171.hg.1 | ENST00000528780.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11000171.hg.1 | NM_003641.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11000171.hg.1 | TC11000011.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11000165.hg.1 | ENST00000328221.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11000165.hg.1 | ENST00000408968.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11000165.hg.1 | ENST00000452428.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11000165.hg.1 | ENST00000528780.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11000165.hg.1 | NM_003641.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11000165.hg.1 | TC11000011.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11000167.hg.1 | ENST00000328221.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11000167.hg.1 | ENST00000408968.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11000167.hg.1 | ENST00000452428.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11000167.hg.1 | ENST00000528780.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11000167.hg.1 | NM_003641.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11000167.hg.1 | TC11000011.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11000169.hg.1 | ENST00000328221.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11000169.hg.1 | ENST00000408968.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11000169.hg.1 | ENST00000452428.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11000169.hg.1 | ENST00000528780.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11000169.hg.1 | NM_003641.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11000169.hg.1 | TC11000011.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012949.hg.1 | ENST00000328221.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012949.hg.1 | ENST00000328221.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012949.hg.1 | ENST00000328221.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012949.hg.1 | ENST00000328221.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012949.hg.1 | ENST00000328221.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012949.hg.1 | ENST00000328221.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012949.hg.1 | ENST00000328221.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012949.hg.1 | ENST00000408968.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012949.hg.1 | ENST00000408968.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012949.hg.1 | ENST00000408968.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012949.hg.1 | ENST00000408968.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012949.hg.1 | ENST00000408968.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012949.hg.1 | ENST00000408968.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012949.hg.1 | ENST00000408968.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012949.hg.1 | ENST00000408968.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012949.hg.1 | ENST00000525554.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012949.hg.1 | ENST00000525554.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012949.hg.1 | ENST00000528780.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012949.hg.1 | ENST00000528780.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012949.hg.1 | ENST00000528780.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012949.hg.1 | ENST00000528780.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012949.hg.1 | ENST00000528780.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012949.hg.1 | ENST00000528780.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012949.hg.1 | NM_003641.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012949.hg.1 | NM_003641.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012949.hg.1 | NM_003641.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012949.hg.1 | NM_003641.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012949.hg.1 | NM_003641.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012949.hg.1 | NM_003641.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012949.hg.1 | NM_003641.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11000163.hg.1 | ENST00000328221.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11000163.hg.1 | ENST00000408968.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11000163.hg.1 | NM_003641.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11000163.hg.1 | TC11000011.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11000164.hg.1 | ENST00000328221.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11000164.hg.1 | ENST00000408968.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11000164.hg.1 | ENST00000452428.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11000164.hg.1 | NM_003641.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11000164.hg.1 | TC11000011.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1100199314.hg.1 | ENST00000328221.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199314.hg.1 | ENST00000408968.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199314.hg.1 | NM_003641.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100199314.hg.1 | ENST00000328221.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199314.hg.1 | ENST00000408968.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199314.hg.1 | NM_003641.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100199314.hg.1 | TC1100012949.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100199315.hg.1 | ENST00000328221.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199315.hg.1 | ENST00000408968.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199315.hg.1 | NM_003641.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100199315.hg.1 | ENST00000328221.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199315.hg.1 | ENST00000408968.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199315.hg.1 | NM_003641.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100199315.hg.1 | TC1100012949.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100199317.hg.1 | ENST00000328221.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199317.hg.1 | ENST00000408968.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199317.hg.1 | ENST00000528780.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199317.hg.1 | NM_003641.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100199317.hg.1 | ENST00000328221.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199317.hg.1 | ENST00000408968.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199317.hg.1 | ENST00000528780.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199317.hg.1 | NM_003641.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100199317.hg.1 | TC1100012949.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100199319.hg.1 | ENST00000328221.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199319.hg.1 | ENST00000408968.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199319.hg.1 | ENST00000525554.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199319.hg.1 | ENST00000528780.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199319.hg.1 | NM_003641.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100199319.hg.1 | ENST00000328221.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199319.hg.1 | ENST00000408968.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199319.hg.1 | ENST00000525554.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199319.hg.1 | ENST00000528780.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199319.hg.1 | NM_003641.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100199319.hg.1 | TC1100012949.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100012949.hg.1 | ENST00000328221.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012949.hg.1 | ENST00000328221.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012949.hg.1 | ENST00000328221.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012949.hg.1 | ENST00000328221.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012949.hg.1 | ENST00000328221.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012949.hg.1 | ENST00000328221.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012949.hg.1 | ENST00000328221.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012949.hg.1 | ENST00000408968.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012949.hg.1 | ENST00000408968.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012949.hg.1 | ENST00000408968.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012949.hg.1 | ENST00000408968.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012949.hg.1 | ENST00000408968.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012949.hg.1 | ENST00000408968.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012949.hg.1 | ENST00000408968.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012949.hg.1 | ENST00000408968.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012949.hg.1 | ENST00000525554.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012949.hg.1 | ENST00000525554.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012949.hg.1 | ENST00000528780.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012949.hg.1 | ENST00000528780.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012949.hg.1 | ENST00000528780.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012949.hg.1 | ENST00000528780.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012949.hg.1 | ENST00000528780.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012949.hg.1 | ENST00000528780.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012949.hg.1 | NM_003641.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012949.hg.1 | NM_003641.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012949.hg.1 | NM_003641.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012949.hg.1 | NM_003641.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012949.hg.1 | NM_003641.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012949.hg.1 | NM_003641.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012949.hg.1 | NM_003641.3 | Refseq |

IFITM1
Genomic Map