Assay Details
| Gene Symbol: RAB11FIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002814.2 | NP_001002814.2 | 4 - 5 | 3577 | 121 | |
| GenBank mRNA | AK090437.1 | - | 1 - 2 | 1266 | 121 | |
| AK122583.1 | - | 1 - 2 | 1716 | 121 | ||
| AK226111.1 | - | 4 - 5 | 3267 | 121 | ||
| AL832155.1 | - | 1 - 2 | 1158 | 121 | ||
| AL833169.1 | - | 1 - 2 | 986 | 121 | ||
| AY280966.1 | - | 1 - 2 | 1998 | 121 | ||
| AY280968.1 | - | 4 - 5 | 3534 | 121 | ||
| AY367050.1 | - | 1 - 2 | 1814 | 121 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000287263.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000287263.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000287263.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000287263.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000287263.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000287263.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000287263.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000287263.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000287263.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000287263.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000330843.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000330843.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000330843.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000330843.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000330843.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000330843.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000330843.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000330843.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000330843.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000330843.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000522727.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000522727.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000522727.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000522727.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000522727.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000522727.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000522774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000522774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000522774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000522774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000522774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000523182.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000523182.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000523182.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000523182.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000524118.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000524118.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000524118.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000524118.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | ENST00000524118.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | NM_001002814.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | NM_001002814.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | NM_001002814.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | NM_001002814.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | NM_001002814.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | NM_001002814.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | NM_001002814.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | NM_001002814.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | NM_001002814.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | NM_001002814.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | NM_001002814.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | NM_025151.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | NM_025151.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | NM_025151.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | NM_025151.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | NM_025151.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | NM_025151.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | NM_025151.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | NM_025151.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | NM_025151.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010138.hg.1 | NM_025151.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000287263.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000287263.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000287263.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000287263.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000287263.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000287263.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000287263.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000287263.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000287263.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000287263.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000330843.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000330843.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000330843.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000330843.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000330843.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000330843.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000330843.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000330843.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000330843.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000330843.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000522727.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000522727.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000522727.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000522727.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000522727.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000522727.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000522774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000522774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000522774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000522774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000522774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000523182.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000523182.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000523182.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000523182.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000524118.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000524118.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000524118.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000524118.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | ENST00000524118.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | NM_001002814.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | NM_001002814.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | NM_001002814.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | NM_001002814.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | NM_001002814.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | NM_001002814.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | NM_001002814.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | NM_001002814.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | NM_001002814.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | NM_001002814.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | NM_001002814.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | NM_025151.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | NM_025151.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | NM_025151.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | NM_025151.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | NM_025151.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | NM_025151.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | NM_025151.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | NM_025151.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | NM_025151.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010138.hg.1 | NM_025151.4 | Refseq |

RAB11FIP1
Genomic Map