Assay Details
| Gene Symbol: KRT7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC042076.1 | - | 1 - 2 | 265 | 114 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 592 | LINC00592 | -21.2kb | Anti-sense | Chr. 12 - 52210930 - 52223813 on NCBI Build GRCh38 | |
| Downstream | keratin 87 pseudogene | KRT87P |
HBD, KRT121P, KRTHBP4 |
4.6kb | Sense | Chr. 12 - 52252026 - 52258521 on NCBI Build GRCh38 |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000546856.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000546856.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000546856.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000546856.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000546856.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000546856.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000546856.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000547613.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000547613.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000547613.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000547613.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000547613.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000547613.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000547613.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000548088.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000548088.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000548088.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000548088.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000548088.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000548657.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000548657.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000548657.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000548657.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000549127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000549127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000549127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000549127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000549638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000549638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000549638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000549638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000549638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000549638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000550153.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000550153.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000550153.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000550153.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000550153.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000550153.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000551130.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000551130.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000551130.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000551130.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000551130.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000552183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000552183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000552183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000552183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000552322.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000552322.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000552322.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000552322.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000552322.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000552322.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000552322.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000552322.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000552322.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000552322.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010744.hg.1 | ENST00000546686.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010744.hg.1 | ENST00000546686.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010744.hg.1 | ENST00000546686.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010744.hg.1 | ENST00000546686.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000546856.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000546856.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000546856.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000546856.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000546856.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000546856.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000546856.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000547613.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000547613.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000547613.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000547613.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000547613.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000547613.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000547613.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000548088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000548088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000548088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000548088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000548088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000548657.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000548657.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000548657.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000548657.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000549127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000549127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000549127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000549127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000549638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000549638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000549638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000549638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000549638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000549638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000550153.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000550153.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000550153.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000550153.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000550153.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000550153.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000551130.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000551130.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000551130.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000551130.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000551130.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000552183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000552183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000552183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000552183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000552322.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000552322.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000552322.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000552322.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000552322.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000552322.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000552322.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000552322.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000552322.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000552322.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | NM_005556.3 | Refseq |

KRT7
Genomic Map