Assay Details
Gene Symbol: ZNF780B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005258595.3 | XP_005258652.1 | 1 - 2 | 81 | 124 | |
XM_017026427.1 | XP_016881916.1 | 1 - 2 | 81 | 124 | ||
GenBank mRNA | AK127063.1 | - | 1 - 2 | 103 | 263 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005258595.3, XM_017026427.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | ENST00000221355.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | ENST00000221355.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | ENST00000221355.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | ENST00000221355.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | ENST00000221355.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | ENST00000221355.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | ENST00000221355.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | ENST00000221355.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | ENST00000221355.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | ENST00000434248.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | ENST00000434248.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | ENST00000434248.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | ENST00000434248.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | ENST00000434248.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | ENST00000434248.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | ENST00000434248.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | ENST00000434248.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | ENST00000434248.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | ENST00000593605.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | ENST00000593605.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | ENST00000593605.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | ENST00000593605.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | ENST00000593605.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | ENST00000593605.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | ENST00000594811.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | ENST00000594811.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | ENST00000594811.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | ENST00000594811.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | ENST00000594811.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | ENST00000594811.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | ENST00000595995.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | ENST00000595995.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | ENST00000595995.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | ENST00000595995.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | ENST00000595995.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | ENST00000598845.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | ENST00000598845.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | ENST00000598845.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | ENST00000598845.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | ENST00000598845.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | ENST00000617676.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | ENST00000617676.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | ENST00000617676.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | ENST00000617676.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | ENST00000617676.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | ENST00000617676.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | ENST00000617676.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | ENST00000617676.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | ENST00000617676.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | NM_001005851.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | NM_001005851.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | NM_001005851.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | NM_001005851.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | NM_001005851.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | NM_001005851.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | NM_001005851.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | NM_001005851.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011951.hg.1 | NM_001005851.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | ENST00000221355.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | ENST00000221355.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | ENST00000221355.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | ENST00000221355.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | ENST00000221355.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | ENST00000221355.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | ENST00000221355.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | ENST00000221355.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | ENST00000221355.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | ENST00000434248.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | ENST00000434248.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | ENST00000434248.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | ENST00000434248.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | ENST00000434248.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | ENST00000434248.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | ENST00000434248.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | ENST00000434248.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | ENST00000434248.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | ENST00000593605.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | ENST00000593605.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | ENST00000593605.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | ENST00000593605.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | ENST00000593605.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | ENST00000593605.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | ENST00000594811.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | ENST00000594811.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | ENST00000594811.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | ENST00000594811.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | ENST00000594811.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | ENST00000594811.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | ENST00000595995.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | ENST00000595995.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | ENST00000595995.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | ENST00000595995.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | ENST00000595995.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | ENST00000598845.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | ENST00000598845.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | ENST00000598845.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | ENST00000598845.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | ENST00000598845.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | ENST00000617676.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | ENST00000617676.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | ENST00000617676.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | ENST00000617676.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | ENST00000617676.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | ENST00000617676.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | ENST00000617676.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | ENST00000617676.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | ENST00000617676.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | NM_001005851.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | NM_001005851.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | NM_001005851.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | NM_001005851.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | NM_001005851.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | NM_001005851.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | NM_001005851.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | NM_001005851.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011951.hg.1 | NM_001005851.2 | Refseq |
