Assay Details
Gene Symbol: KRTAP4-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030976.1 | NP_112238.1 | 1 | 967 | 162 | |
GenBank mRNA | AJ406938.1 | - | 1 | 666 | 162 | |
AJ406945.1 | - | 1 | 931 | 162 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | keratin associated protein 4-12 | KRTAP4-12 |
KAP4.12, KRTAP4.12 |
-15.3kb | Sense | Chr. 17 - 41123091 - 41124167 on NCBI Build GRCh38 |
Downstream | keratin associated protein 4-5 | KRTAP4-5 |
KAP4.5, KRTAP4.5 |
8.4kb | Sense | Chr. 17 - 41148924 - 41149802 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC17001491.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
