Assay Details
| Gene Symbol: HSD17B12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016142.2 | NP_057226.1 | 1 - 2 | 397 | 132 | |
| GenBank mRNA | AF078850.1 | - | 1 - 2 | 132 | ||
| AK027882.1 | - | 291 | 132 | |||
| AK074952.1 | - | 1 - 2 | 249 | 132 | ||
| AK075011.1 | - | 1 - 2 | 282 | 132 | ||
| AK075216.1 | - | 1 - 2 | 258 | 132 | ||
| AK222881.1 | - | 1 - 2 | 132 | |||
| AK292625.1 | - | 1 - 2 | 282 | 132 | ||
| AK301663.1 | - | 1 - 3 | 108 | |||
| BC012043.1 | - | 1 - 2 | 220 | 3622879 | 132 | |
| BC012536.1 | - | 1 - 2 | 213 | 132 | ||
More Information
Set Membership: |
|

HSD17B12
Genomic Map