Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006766.hg.1 |
ENST00000390287.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006766.hg.1 |
ENST00000390287.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008113.hg.1 |
ENST00000337471.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008113.hg.1 |
ENST00000337471.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008113.hg.1 |
ENST00000337471.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008113.hg.1 |
ENST00000337471.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008113.hg.1 |
ENST00000337471.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008113.hg.1 |
ENST00000337471.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008113.hg.1 |
ENST00000337471.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008113.hg.1 |
ENST00000337471.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008113.hg.1 |
ENST00000337471.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008113.hg.1 |
ENST00000337471.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008113.hg.1 |
ENST00000337471.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008113.hg.1 |
ENST00000337471.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008113.hg.1 |
ENST00000419303.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008113.hg.1 |
ENST00000419303.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008113.hg.1 |
ENST00000419303.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008113.hg.1 |
ENST00000419303.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008113.hg.1 |
ENST00000419303.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008113.hg.1 |
ENST00000419303.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008113.hg.1 |
ENST00000419303.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008113.hg.1 |
ENST00000419303.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008113.hg.1 |
ENST00000548391.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008113.hg.1 |
ENST00000548391.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008113.hg.1 |
ENST00000548391.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008113.hg.1 |
ENST00000548391.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008113.hg.1 |
ENST00000548391.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006766.hg.1 |
ENST00000390287.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006766.hg.1 |
ENST00000390287.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008113.hg.1 |
ENST00000337471.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008113.hg.1 |
ENST00000337471.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008113.hg.1 |
ENST00000337471.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008113.hg.1 |
ENST00000337471.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008113.hg.1 |
ENST00000337471.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008113.hg.1 |
ENST00000337471.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008113.hg.1 |
ENST00000337471.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008113.hg.1 |
ENST00000337471.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008113.hg.1 |
ENST00000337471.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008113.hg.1 |
ENST00000337471.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008113.hg.1 |
ENST00000337471.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008113.hg.1 |
ENST00000337471.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008113.hg.1 |
ENST00000419303.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008113.hg.1 |
ENST00000419303.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008113.hg.1 |
ENST00000419303.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008113.hg.1 |
ENST00000419303.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008113.hg.1 |
ENST00000419303.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008113.hg.1 |
ENST00000419303.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008113.hg.1 |
ENST00000419303.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008113.hg.1 |
ENST00000419303.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008113.hg.1 |
ENST00000548391.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008113.hg.1 |
ENST00000548391.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008113.hg.1 |
ENST00000548391.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008113.hg.1 |
ENST00000548391.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008113.hg.1 |
ENST00000548391.1 |
Ensembl |