Assay Details
Gene Symbol: FAM109A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK093052.1 | - | 1 - 2 | 74 | 89 | |
DB055268.1 | - | 1 - 2 | 74 | 89 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK093052.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | ENST00000361483.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | ENST00000361483.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | ENST00000361483.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | ENST00000361483.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | ENST00000361483.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | ENST00000361483.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | ENST00000361483.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | ENST00000361483.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | ENST00000361483.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | ENST00000361483.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | ENST00000361483.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | ENST00000547710.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | ENST00000547710.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | ENST00000547710.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | ENST00000547710.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | ENST00000547710.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | ENST00000547838.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | ENST00000547838.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | ENST00000547838.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | ENST00000547838.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | ENST00000547838.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | ENST00000547838.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | ENST00000547838.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | ENST00000547838.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | ENST00000548163.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | ENST00000548163.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | ENST00000548163.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | ENST00000548163.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | ENST00000548163.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | ENST00000548163.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | ENST00000548163.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | ENST00000549321.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | ENST00000549321.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | ENST00000549321.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | ENST00000549321.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | ENST00000549321.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | NM_001177996.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | NM_001177996.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | NM_001177996.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | NM_001177996.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | NM_001177996.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | NM_001177996.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | NM_001177996.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | NM_001177996.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | NM_001177996.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | NM_001177996.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | NM_001177996.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | NM_001177997.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | NM_001177997.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | NM_001177997.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | NM_001177997.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | NM_001177997.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | NM_001177997.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | NM_001177997.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | NM_001177997.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | NM_001177997.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | NM_001177997.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | NM_144671.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | NM_144671.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | NM_144671.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | NM_144671.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | NM_144671.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | NM_144671.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | NM_144671.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | NM_144671.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | NM_144671.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011931.hg.1 | NM_144671.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | ENST00000361483.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | ENST00000361483.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | ENST00000361483.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | ENST00000361483.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | ENST00000361483.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | ENST00000361483.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | ENST00000361483.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | ENST00000361483.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | ENST00000361483.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | ENST00000361483.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | ENST00000361483.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | ENST00000547710.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | ENST00000547710.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | ENST00000547710.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | ENST00000547710.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | ENST00000547710.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | ENST00000547838.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | ENST00000547838.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | ENST00000547838.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | ENST00000547838.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | ENST00000547838.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | ENST00000547838.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | ENST00000547838.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | ENST00000547838.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | ENST00000548163.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | ENST00000548163.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | ENST00000548163.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | ENST00000548163.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | ENST00000548163.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | ENST00000548163.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | ENST00000548163.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | ENST00000549321.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | ENST00000549321.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | ENST00000549321.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | ENST00000549321.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | ENST00000549321.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | NM_001177996.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | NM_001177996.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | NM_001177996.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | NM_001177996.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | NM_001177996.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | NM_001177996.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | NM_001177996.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | NM_001177996.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | NM_001177996.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | NM_001177996.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | NM_001177996.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | NM_001177997.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | NM_001177997.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | NM_001177997.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | NM_001177997.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | NM_001177997.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | NM_001177997.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | NM_001177997.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | NM_001177997.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | NM_001177997.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | NM_001177997.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | NM_144671.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | NM_144671.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | NM_144671.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | NM_144671.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | NM_144671.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | NM_144671.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | NM_144671.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | NM_144671.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | NM_144671.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011931.hg.1 | NM_144671.4 | Refseq |
