Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011668.hg.1 |
ENST00000431347.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011668.hg.1 |
ENST00000431347.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011668.hg.1 |
ENST00000431347.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011668.hg.1 |
ENST00000431347.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011668.hg.1 |
ENST00000431347.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011668.hg.1 |
NR_110797.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011668.hg.1 |
NR_110797.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011668.hg.1 |
NR_110797.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011668.hg.1 |
NR_110797.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011668.hg.1 |
NR_110797.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011668.hg.1 |
ENST00000431347.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011668.hg.1 |
ENST00000431347.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011668.hg.1 |
ENST00000431347.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011668.hg.1 |
ENST00000431347.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011668.hg.1 |
ENST00000431347.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011668.hg.1 |
NR_110797.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011668.hg.1 |
NR_110797.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011668.hg.1 |
NR_110797.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011668.hg.1 |
NR_110797.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011668.hg.1 |
NR_110797.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100185905.hg.1 |
ENST00000431347.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185905.hg.1 |
NR_110797.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100185905.hg.1 |
ENST00000431347.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185905.hg.1 |
NR_110797.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100185905.hg.1 |
TC0100011668.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100185906.hg.1 |
ENST00000431347.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185906.hg.1 |
NR_110797.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100185906.hg.1 |
ENST00000431347.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185906.hg.1 |
NR_110797.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100185906.hg.1 |
TC0100011668.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100185904.hg.1 |
ENST00000431347.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185904.hg.1 |
NR_110797.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100185904.hg.1 |
ENST00000431347.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185904.hg.1 |
NR_110797.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100185904.hg.1 |
TC0100011668.hg.1 |
Clariom D TC |