Assay Details
Gene Symbol: EIF3IP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_003024.1 | 1 | 932 | 80 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | chromosome 7 open reading frame 66 | C7orf66 | -1.1mb | Sense | Chr. 7 - 108883975 - 108884587 on NCBI Build GRCh38 | |
Downstream | inner mitochondrial membrane peptidase subunit 2 | IMMP2L |
IMMP2L-IT1, IMP2, IMP2-LIKE |
702.4kb | Sense | Chr. 7 - 110662645 - 111562531 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
ArrayStar
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3412563 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4056 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4057 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 6626 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3412563 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3412563 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25942016 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25941999 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05685187 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25942010 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05685202 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05685208 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25942009 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05685189 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05685201 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25942014 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05685183 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05685188 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25942001 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25941974 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05685212 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05685185 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25942005 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05685200 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05685180 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25942007 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25942013 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05685209 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25942011 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25941984 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25942015 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05685194 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25942002 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16004515 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25941997 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25941991 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05685193 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16004515 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05685202 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25942016 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25942010 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05685193 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25942014 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25941997 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05685189 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05685194 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25942015 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05685208 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05685212 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25942009 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25941984 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05685201 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25942011 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25942013 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25942005 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05685185 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05685188 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25941999 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05685180 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05685200 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25942002 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05685183 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25942007 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25941991 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05685209 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25941974 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05685187 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25942001 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05685160 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05685166 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16004517 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05685170 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05685205 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05685209 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05685181 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16004522 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05685175 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16004514 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05685183 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05685190 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05685202 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05685212 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05685216 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05685193 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05685188 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05685220 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05685169 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16004519 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05685197 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05685164 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05685180 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05685179 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05685167 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05685174 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05685201 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05685185 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16004520 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05685191 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05685222 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A033377 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A033377 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 167643 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1809516 | NR_003024.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1809516 | NR_003024.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1809516 | NR_003024.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1809516 | NR_003024.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1809516 | NR_003024.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1809516 | NR_003024.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1809516 | NR_003024.1 | human_transcriptome |
