Assay Details
| Gene Symbol: HAVCR1P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003603.1 | 1 | 81 | |||
| GenBank mRNA | BX114328.1 | - | 1 | 185 | 81 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 254 | ZNF254 |
BMZF-5, HD-ZNF1, ZNF539, ZNF91L |
-32.8kb | Sense | Chr. 19 - 24033405 - 24129968 on NCBI Build GRCh38 |
| Downstream | endogenous retrovirus group K member 19 Pol protein | LOC105376906 | 3.4mb | Anti-sense | Chr. 19 - 27520104 - 27641535 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001374.hg.1
Set Membership: |
|

HAVCR1P1
Genomic Map