Assay Details
Gene Symbol: ZNF138 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_073389.1 | 3 | 1417 | 149 | ||
GenBank mRNA | AL834121.1 | - | 1 | 149 | ||
BC017945.1 | - | 1 | 277 | 149 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
