Assay Details
| Gene Symbol: DPY19L2P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003561.2 | 3 - 4 | 343 | 151 | ||
| GenBank mRNA | AK302046.1 | - | 2 - 3 | 551 | 151 | |
| BC023575.2 | - | 2 - 3 | 740 | 151 | ||
More Information
Updated Mapping Information
Set Membership: |
|

DPY19L2P2
Genomic Map