Assay Details
| Gene Symbol: SCD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005063.4 | NP_005054.3 | 2 - 3 | 798 | 129 | |
| GenBank mRNA | AB032261.1 | - | 2 - 3 | 688 | 129 | |
| AB208982.1 | - | 2 - 3 | 555 | 129 | ||
| AF097514.1 | - | 2 - 3 | 543 | 129 | ||
| AK222862.1 | - | 2 - 3 | 515 | 129 | ||
| AK312312.1 | - | 2 - 3 | 580 | 129 | ||
| BC005807.2 | - | 2 - 3 | 485 | 129 | ||
| BC062303.1 | - | 2 - 3 | 684 | 6586153 | 129 | |
| S70284.1 | - | 1 - 2 | 303 | 129 | ||
| Y13647.1 | - | 2 - 3 | 516 | 129 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000721.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10008386.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008386.hg.1 | TC10000721.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008389.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008389.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008389.hg.1 | TC10000721.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008390.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008390.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008390.hg.1 | TC10000721.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008394.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008394.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008394.hg.1 | TC10000721.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008392.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008392.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008392.hg.1 | TC10000721.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008396.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008396.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008396.hg.1 | TC10000721.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008397.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008397.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008397.hg.1 | TC10000721.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008393.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008393.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008393.hg.1 | TC10000721.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008643.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008643.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008643.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008643.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008643.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008643.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008643.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008643.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008643.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008643.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008643.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008643.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008643.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008643.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008643.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008643.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008643.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008643.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008643.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008643.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008643.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008643.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008643.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008643.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008643.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007240.hg.1 | ENST00000455244.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008391.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008391.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008391.hg.1 | TC10000721.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008398.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008398.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008398.hg.1 | TC10000721.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006916.hg.1 | ENST00000502928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008395.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008395.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008395.hg.1 | TC10000721.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1000158932.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000158932.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000158932.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000158932.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000158932.hg.1 | TC1000008643.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000158933.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000158933.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000158933.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000158933.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000158933.hg.1 | TC1000008643.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000158935.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000158935.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000158935.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000158935.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000158935.hg.1 | TC1000008643.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000158927.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000158927.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000158927.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000158927.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000158927.hg.1 | TC1000008643.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000158929.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000158929.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000158929.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000158929.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000158929.hg.1 | TC1000008643.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000158926.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000158926.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000158926.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000158926.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000158926.hg.1 | TC1000008643.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000158931.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000158931.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000158931.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000158931.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000158931.hg.1 | TC1000008643.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500006916.hg.1 | ENST00000502928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000158928.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000158928.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000158928.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000158928.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000158928.hg.1 | TC1000008643.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000158934.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000158934.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000158934.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000158934.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000158934.hg.1 | TC1000008643.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700007240.hg.1 | ENST00000455244.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000158924.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000158924.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000158924.hg.1 | TC1000008643.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000158930.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000158930.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000158930.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000158930.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000158930.hg.1 | TC1000008643.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000008643.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008643.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008643.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008643.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008643.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008643.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008643.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008643.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008643.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008643.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008643.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008643.hg.1 | ENST00000370355.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008643.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008643.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008643.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008643.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008643.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008643.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008643.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008643.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008643.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008643.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008643.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008643.hg.1 | NM_005063.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008643.hg.1 | NM_005063.4 | Refseq |

SCD
Genomic Map