Assay Details
| Gene Symbol: CTAGE10P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003268.3 | 1 | 148 | 124 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | karyopherin subunit alpha 3 | KPNA3 |
IPOA4, SRP1, SRP1gamma, SRP4, hSRP1 |
-97.5kb | Anti-sense | Chr. 13 - 49699307 - 49792921 on NCBI Build GRCh38 |
| Downstream | SPRY domain containing 7 | SPRYD7 |
C13orf1, CLLD6 |
19.3kb | Anti-sense | Chr. 13 - 49912702 - 49936501 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC13000201.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08842394 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29290330 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08842397 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29290327 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08842387 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29290325 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08842380 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29290329 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08842404 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29290342 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29290338 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29290333 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08842406 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08842405 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29290346 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 307261 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 104416 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 347880 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P358054 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3340007 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P366493 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 37700 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 8513 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 56418 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 48208 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P358054 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3340007 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3340007 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_24_P358054 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08842380 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08842380 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P366493 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P02775460 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P358054 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P708438 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08842381 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08842397 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08842391 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08842410 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08842385 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08842390 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08842401 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08842383 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08842388 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08842395 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08842407 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08842405 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08842403 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08842393 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NR_003268_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7969152 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7969151 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007319.hg.1 | ENST00000435601.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007319.hg.1 | ENST00000435601.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007319.hg.1 | ENST00000435601.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007319.hg.1 | ENST00000435601.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13002397.hg.1 | NR_003268.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13002397.hg.1 | TC13000201.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007178.hg.1 | ENST00000318353.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007178.hg.1 | NR_003268.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007178.hg.1 | NR_003268.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007178.hg.1 | NR_003268.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300148641.hg.1 | ENST00000318353.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300148641.hg.1 | NR_003268.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300148641.hg.1 | ENST00000318353.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300148641.hg.1 | NR_003268.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300148641.hg.1 | TC1300007178.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1300007319.hg.1 | ENST00000435601.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007319.hg.1 | ENST00000435601.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007319.hg.1 | ENST00000435601.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007319.hg.1 | ENST00000435601.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300148640.hg.1 | NR_003268.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300148640.hg.1 | NR_003268.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300148640.hg.1 | TC1300007178.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1300148642.hg.1 | NR_003268.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300148642.hg.1 | NR_003268.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300148642.hg.1 | TC1300007178.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1300007178.hg.1 | ENST00000318353.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007178.hg.1 | NR_003268.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007178.hg.1 | NR_003268.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007178.hg.1 | NR_003268.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A050182 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A050181 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A050182 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A050181 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 104523 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_2193195 | BI463404.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1654061 | - | - |
| HumanHT-12 V4.0 expression beadchip | ILMN_2193195 | BI463404.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2193195 | BI463404.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2193195 | BI463404.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1654061 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2193195 | BI463404.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2193195 | BI463404.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2193195 | BI463404.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1654061 | - | - |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2193195 | BI463404.1 | human_transcriptome |

CTAGE10P
Genomic Map