Assay Details
| Gene Symbol: AUTS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127231.2 | NP_001120703.1 | 2 - 3 | 1260 | 72 | |
| NM_001127232.2 | NP_001120704.1 | 2 - 3 | 1260 | 72 | ||
| NM_015570.3 | NP_056385.1 | 2 - 3 | 1260 | 72 | ||
| XM_011516010.2 | XP_011514312.1 | 2 - 3 | 1704 | 72 | ||
| XM_011516011.2 | XP_011514313.1 | 2 - 3 | 1704 | 72 | ||
| XM_011516012.2 | XP_011514314.1 | 2 - 3 | 1704 | 72 | ||
| XM_011516013.2 | XP_011514315.1 | 2 - 3 | 1704 | 72 | ||
| XM_011516014.2 | XP_011514316.1 | 2 - 3 | 1704 | 72 | ||
| XM_011516018.2 | XP_011514320.1 | 2 - 3 | 239 | 72 | ||
| XM_017011951.1 | XP_016867440.1 | 2 - 3 | 1704 | 72 | ||
| GenBank mRNA | AF326917.1 | - | 2 - 3 | 846 | 72 | |
| BC011643.2 | - | 2 - 3 | 568 | 72 | ||
| BC064693.1 | - | 2 - 3 | 1268 | 30345831 | 72 | |
| JQ670866.1 | - | 2 - 3 | 525 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011516018.2 transcripts.
Set Membership: |
|

AUTS2
Genomic Map