Assay Details
| Gene Symbol: NR2F2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BI758289.1 | - | 449 | 86 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NR2F2-AS1| Gene Symbol: NR2F2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BI758289.1 | - | 449 | 86 | ||