| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X014333.hg.1 |
TC0X001063.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X014333.hg.1 |
ENST00000462054.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X014333.hg.1 |
NM_001243197.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X014334.hg.1 |
TC0X001063.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X014334.hg.1 |
ENST00000462054.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X014334.hg.1 |
NM_001243197.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000462054.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000462054.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000462054.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000462054.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000462054.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000462054.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000485377.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000485377.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000485377.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000485377.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000485377.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000498281.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000498281.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000498281.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000498281.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
ENST00000498281.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_001243197.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_001243197.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_001243197.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_001243197.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_001243197.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_001243197.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X014335.hg.1 |
TC0X001063.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X014335.hg.1 |
ENST00000462054.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X014335.hg.1 |
NM_001243197.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000375365.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000396435.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000462054.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000462054.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000462054.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000462054.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000462054.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000462054.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000485377.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000485377.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000485377.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000485377.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000485377.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000498281.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000498281.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000498281.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000498281.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
ENST00000498281.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_001111125.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_001243197.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_001243197.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_001243197.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_001243197.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_001243197.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_001243197.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009755.hg.1 |
NM_015075.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00166645.hg.1 |
ENST00000462054.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00166645.hg.1 |
ENST00000485377.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00166645.hg.1 |
NM_001243197.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00166645.hg.1 |
ENST00000462054.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00166645.hg.1 |
ENST00000485377.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00166645.hg.1 |
NM_001243197.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00166645.hg.1 |
TC0X00009755.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00166644.hg.1 |
ENST00000462054.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00166644.hg.1 |
ENST00000485377.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00166644.hg.1 |
ENST00000498281.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00166644.hg.1 |
NM_001243197.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00166644.hg.1 |
ENST00000462054.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00166644.hg.1 |
ENST00000485377.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00166644.hg.1 |
ENST00000498281.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00166644.hg.1 |
NM_001243197.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00166644.hg.1 |
TC0X00009755.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00166646.hg.1 |
ENST00000462054.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00166646.hg.1 |
ENST00000485377.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00166646.hg.1 |
ENST00000498281.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00166646.hg.1 |
NM_001243197.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00166646.hg.1 |
ENST00000462054.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00166646.hg.1 |
ENST00000485377.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00166646.hg.1 |
ENST00000498281.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00166646.hg.1 |
NM_001243197.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00166646.hg.1 |
TC0X00009755.hg.1 |
Clariom D TC |