Assay Details
| Gene Symbol: LOC101927434 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_125943.1 | 2 | 82 | 110 | ||
| GenBank mRNA | BX092247.1 | - | 2 | 82 | 110 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927434| Gene Symbol: LOC101927434 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_125943.1 | 2 | 82 | 110 | ||
| GenBank mRNA | BX092247.1 | - | 2 | 82 | 110 | |
Set Membership: |
|