Assay Details
| Gene Symbol: TMEM14EP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_132656.1 | 1 | 626 | 120 | ||
| GenBank mRNA | AW866456.1 | - | 1 | 353 | 120 | |
| AY358260.1 | - | 1 | 625 | 120 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P932416 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P932416 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 6224 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 47191 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P932416 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_24_P932416 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02680116 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820279 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820287 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820256 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15501059 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02680152 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820264 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820277 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820259 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02680092 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820307 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15501056 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820263 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820249 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820290 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820298 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02680140 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02680159 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02680129 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820243 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820253 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820254 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820244 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820240 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820276 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820282 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820304 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820286 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820291 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820268 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820250 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02680141 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02680134 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820274 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820293 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820306 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02680123 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02680095 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02680157 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02680111 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820246 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820305 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820245 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820297 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820302 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820261 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820242 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820280 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820237 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820272 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820234 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02680145 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820262 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820294 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820270 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820253 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820262 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820234 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820268 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15501059 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820237 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820256 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820282 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02680140 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820297 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820280 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820286 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820261 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820250 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02680141 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820274 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820298 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820245 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820277 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820304 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820294 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820307 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820240 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02680111 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820306 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02680134 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820291 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820254 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02680092 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820246 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820279 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02680145 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820259 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02680152 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02680159 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02680123 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820287 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02680129 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15501056 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820305 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02680157 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820270 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820276 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820263 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820302 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820243 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820293 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02680095 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820290 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820264 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820244 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820242 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820249 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22820272 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02680116 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P932416 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P932416 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02680134 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02680139 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15501062 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02680158 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02680151 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02680162 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02680107 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15501058 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02680091 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02680169 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02680100 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02680110 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02680095 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02680144 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02680120 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02680104 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02680115 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02680150 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15501064 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02680122 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02680143 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02680156 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02680168 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02680112 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15501056 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02680173 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02680093 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02680165 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02680127 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02680147 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02680137 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02680132 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02680153 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02680102 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02680141 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02680124 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02680171 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02680130 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02680136 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02680126 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02680175 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NM_001123228_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8091548 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8091547 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8091549 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8091546 | - | - |
| Affymetrix Human Genome U219 Array | 11738565_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11738565_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A021787 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A021787 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3236468 | AY358260.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3236468 | NM_001123228.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3236468 | AY358260.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3236468 | NM_001123228.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3236468 | AY358260.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3236468 | NM_001123228.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3236468 | AY358260.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3236468 | NM_001123228.1 | human_transcriptome |

TMEM14EP
Genomic Map