Assay Details
| Gene Symbol: TOP1P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_001283.1 | 1 | 371 | 103 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
POM121 transmembrane nucleoporin like 10, pseudogene |
POM121L10P | -105.4kb | Anti-sense | Chr. 22 - 24645166 - 24659147 on NCBI Build GRCh38 | |
| Downstream | small G protein signaling modulator 1 | SGSM1 | RUTBC2 | 40.2kb | Sense | Chr. 22 - 24806221 - 24926846 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000157.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P374382 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P374382 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 1028 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 45696 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P305507 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P374382 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_24_P374382 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11447509 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11447526 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32126649 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32126644 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17269592 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11447513 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11447551 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32126673 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11447552 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32126634 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32126676 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11447522 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32126638 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32126656 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11447550 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17269589 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32126632 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11447521 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11447537 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11447520 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11447542 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17269590 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32126659 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32126674 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11447547 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32126646 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32126678 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11447511 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11447549 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32126669 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11447553 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32126640 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11447518 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11447516 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32126643 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11447555 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11447538 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32126653 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32126629 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32126639 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11447540 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32126647 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11447526 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32126643 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32126669 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32126639 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11447522 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17269590 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11447520 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32126634 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32126629 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32126673 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32126632 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11447540 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11447521 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11447552 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32126646 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11447537 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11447513 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32126678 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32126653 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11447511 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11447553 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32126638 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11447542 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32126649 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11447550 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32126644 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17269589 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32126656 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11447509 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11447555 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17269592 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11447549 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11447538 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32126676 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11447518 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32126647 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11447547 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32126640 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11447516 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32126674 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11447551 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32126659 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P374382 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P374382 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11447535 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11447539 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17269611 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11447568 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11447572 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11447540 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11447510 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11447549 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11447557 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11447532 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11447567 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11447544 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11447522 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11447550 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17269610 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11447530 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11447524 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11447517 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11447515 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11447512 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11447554 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11447505 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11447547 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11447548 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11447573 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11447527 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11447508 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17269609 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11447552 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11447519 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11447537 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11447542 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11447503 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 1566833_PM_x_at | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1566832_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR22002962.hg.1 | NR_001283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22002962.hg.1 | TC22000157.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2200147909.hg.1 | TC2200006906.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200147909.hg.1 | NR_001283.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200147909.hg.1 | NR_001283.1 | Refseq |
| Human Genome U133 Plus 2.0 Array | 1566832_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1566832_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1566833_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1566833_PM_x_at | - | - |
| Human Genome U95 Set | 31680_PM_at | - | - |
| Human Genome U95 Set | 31680_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566832_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566833_x_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566832_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566833_x_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566832_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566832_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566833_x_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566832_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566832_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566832_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566832_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566833_x_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566833_x_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566832_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566833_x_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566833_x_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566833_x_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566833_x_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566833_x_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566832_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566832_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566833_x_at6 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A020243 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A020243 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 236556 | - | - |
| ABI Human Genome Survey Microarray Version 2.0 | 236677 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_2043109 | NR_001283.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2043109 | NR_001283.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2043109 | NR_001283.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2043109 | NR_001283.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2043109 | NR_001283.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2043109 | NR_001283.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2043109 | NR_001283.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2043109 | NR_001283.1 | human_transcriptome |

TOP1P2
Genomic Map