Assay Details
Gene Symbol: LOC101928673 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_125960.1 | 2 | 1476 | 110 | ||
GenBank mRNA | AK130460.1 | - | 2 | 1477 | 110 | |
BC136808.1 | - | 2 | 1469 | 110 | ||
BU735514.1 | - | 1 | 110 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603212 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662492 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603307 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603246 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662586 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603303 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662574 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603275 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603254 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662563 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603302 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603315 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603264 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603187 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662460 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603199 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662539 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603201 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662465 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603192 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662502 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603277 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603218 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603190 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603248 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603204 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662489 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603280 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662495 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662553 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603281 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603186 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603200 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662511 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662594 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662578 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662582 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662541 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603170 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662514 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603259 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603214 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603286 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603238 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662487 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603267 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662564 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603189 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603184 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662518 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603240 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662527 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603272 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603278 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603316 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603217 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603174 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662469 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603232 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603245 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603247 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603298 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603230 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662468 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603276 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662559 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603270 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603283 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662477 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662481 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603183 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662587 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662484 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603235 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662551 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603185 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603222 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662517 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603291 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662504 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603169 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662520 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662491 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662562 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15144360 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603285 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662519 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662552 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662575 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662490 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15144367 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662524 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603288 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662466 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662526 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662523 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603265 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662479 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662540 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662513 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662573 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662572 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662589 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603263 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662486 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662515 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662516 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662501 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603219 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603311 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603234 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603178 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603312 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662521 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603309 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603172 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603268 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662593 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603300 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00603279 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20662498 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P52119 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 26141 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 22316 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 7922352 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7922351 | - | - |
Affymetrix Human Clariom D Assay | PSR0100224839.hg.1 | ENST00000367716.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100224839.hg.1 | NR_125960.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100224839.hg.1 | ENST00000367716.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100224839.hg.1 | NR_125960.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100224839.hg.1 | TC0100016432.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100224838.hg.1 | ENST00000367716.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100224838.hg.1 | NR_125960.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100224838.hg.1 | ENST00000367716.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100224838.hg.1 | NR_125960.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100224838.hg.1 | TC0100016432.hg.1 | Clariom D TC |
