| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01009597.hg.1 |
TC01000570.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01009597.hg.1 |
NR_024270.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013999.hg.1 |
ENST00000359600.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013999.hg.1 |
ENST00000359600.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013999.hg.1 |
ENST00000359600.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013999.hg.1 |
ENST00000359600.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013999.hg.1 |
ENST00000359600.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013999.hg.1 |
ENST00000359600.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013999.hg.1 |
ENST00000359600.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013999.hg.1 |
ENST00000359600.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013999.hg.1 |
ENST00000359600.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013999.hg.1 |
ENST00000359600.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013999.hg.1 |
ENST00000359600.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013999.hg.1 |
ENST00000359600.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013999.hg.1 |
ENST00000359600.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013999.hg.1 |
ENST00000359600.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013999.hg.1 |
ENST00000359600.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013999.hg.1 |
ENST00000359600.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013999.hg.1 |
ENST00000359600.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013999.hg.1 |
ENST00000464588.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013999.hg.1 |
ENST00000464588.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013999.hg.1 |
ENST00000464588.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013999.hg.1 |
NM_020883.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013999.hg.1 |
NM_020883.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013999.hg.1 |
NM_020883.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013999.hg.1 |
NM_020883.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013999.hg.1 |
NM_020883.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013999.hg.1 |
NM_020883.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013999.hg.1 |
NM_020883.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013999.hg.1 |
NM_020883.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013999.hg.1 |
NM_020883.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013999.hg.1 |
NM_020883.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013999.hg.1 |
NM_020883.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013999.hg.1 |
NM_020883.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013999.hg.1 |
NM_020883.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013999.hg.1 |
NM_020883.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013999.hg.1 |
NM_020883.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013999.hg.1 |
NM_020883.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013999.hg.1 |
NM_020883.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008118.hg.1 |
ENST00000626198.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008118.hg.1 |
ENST00000626198.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008118.hg.1 |
ENST00000626198.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008118.hg.1 |
NR_024270.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008118.hg.1 |
NR_024270.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008118.hg.1 |
NR_024270.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100157951.hg.1 |
ENST00000626198.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100157951.hg.1 |
NR_024270.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100157951.hg.1 |
ENST00000626198.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100157951.hg.1 |
NR_024270.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100157951.hg.1 |
TC0100008118.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100008118.hg.1 |
ENST00000626198.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008118.hg.1 |
ENST00000626198.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008118.hg.1 |
ENST00000626198.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008118.hg.1 |
NR_024270.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008118.hg.1 |
NR_024270.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008118.hg.1 |
NR_024270.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100157950.hg.1 |
ENST00000626198.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100157950.hg.1 |
NR_024270.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100157950.hg.1 |
ENST00000626198.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100157950.hg.1 |
NR_024270.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100157950.hg.1 |
TC0100008118.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100013999.hg.1 |
ENST00000359600.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013999.hg.1 |
ENST00000359600.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013999.hg.1 |
ENST00000359600.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013999.hg.1 |
ENST00000359600.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013999.hg.1 |
ENST00000359600.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013999.hg.1 |
ENST00000359600.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013999.hg.1 |
ENST00000359600.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013999.hg.1 |
ENST00000359600.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013999.hg.1 |
ENST00000359600.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013999.hg.1 |
ENST00000359600.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013999.hg.1 |
ENST00000359600.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013999.hg.1 |
ENST00000359600.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013999.hg.1 |
ENST00000359600.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013999.hg.1 |
ENST00000359600.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013999.hg.1 |
ENST00000359600.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013999.hg.1 |
ENST00000359600.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013999.hg.1 |
ENST00000359600.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013999.hg.1 |
ENST00000464588.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013999.hg.1 |
ENST00000464588.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013999.hg.1 |
ENST00000464588.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013999.hg.1 |
NM_020883.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013999.hg.1 |
NM_020883.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013999.hg.1 |
NM_020883.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013999.hg.1 |
NM_020883.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013999.hg.1 |
NM_020883.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013999.hg.1 |
NM_020883.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013999.hg.1 |
NM_020883.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013999.hg.1 |
NM_020883.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013999.hg.1 |
NM_020883.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013999.hg.1 |
NM_020883.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013999.hg.1 |
NM_020883.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013999.hg.1 |
NM_020883.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013999.hg.1 |
NM_020883.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013999.hg.1 |
NM_020883.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013999.hg.1 |
NM_020883.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013999.hg.1 |
NM_020883.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013999.hg.1 |
NM_020883.1 |
Refseq |