Assay Details
| Gene Symbol: C1orf145 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_073155.1 | 2 | 780 | 110 | ||
| GenBank mRNA | BC027909.1 | - | 2 | 780 | 110 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

C1orf145
Genomic Map