Assay Details
| Gene Symbol: FAM101B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182705.2 | NP_874364.1 | 2 | 2424 | 90 | |
| GenBank mRNA | AK093557.1 | - | 1 | 1118 | 90 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

FAM101B
Genomic Map