Assay Details
| Gene Symbol: LINC00868 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC038218.1 | - | 1 | 359 | 165 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17018367 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31056917 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31056943 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10472491 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10472470 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31056950 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31056912 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10472465 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10472462 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10472445 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10472480 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10472472 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10472444 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10472493 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10472475 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10472456 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31056945 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10472438 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10472446 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10472487 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10472476 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10472471 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10472437 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10472453 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10472482 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10472488 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10472452 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10472473 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10472490 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31056937 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10472443 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10472466 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31056926 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10472498 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10472494 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10472496 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10472497 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10472468 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10472469 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31056920 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10472457 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31056916 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10472499 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10472461 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10472447 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10472440 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31056929 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31056911 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 399011 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 945849 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 838891 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3839820 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 41888 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3839820 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 91021 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03293247 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03293251 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P20733564 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P20983706 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P794508 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_18_P20983706 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P03293247 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P20733564 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 1556533_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008973.hg.1 | ENST00000589708.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008973.hg.1 | ENST00000589708.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008973.hg.1 | ENST00000589708.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008973.hg.1 | ENST00000589708.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008973.hg.1 | ENST00000589914.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008973.hg.1 | ENST00000589914.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008973.hg.1 | ENST00000589914.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008973.hg.1 | ENST00000592919.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008973.hg.1 | ENST00000592919.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008973.hg.1 | ENST00000589708.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008973.hg.1 | ENST00000589708.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008973.hg.1 | ENST00000589708.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008973.hg.1 | ENST00000589708.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008973.hg.1 | ENST00000589914.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008973.hg.1 | ENST00000589914.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008973.hg.1 | ENST00000589914.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008973.hg.1 | ENST00000592919.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008973.hg.1 | ENST00000592919.1 | Ensembl |
| Human Genome U133 Plus 2.0 Array | 1556533_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1556533_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556533_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556533_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556533_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556533_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556533_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556533_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556533_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556533_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556533_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556533_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556533_at9 | - | - |

LINC00868
Genomic Map