Assay Details
| Gene Symbol: LOC100288798 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_125377.1 | 4 | 71 | |||
| NR_125378.1 | 4 | 71 | ||||
| NR_125379.1 | 4 | 71 | ||||
| NR_125380.1 | 4 | 71 | ||||
| NR_125381.1 | 3 | 71 | ||||
| GenBank mRNA | AI094227.1 | - | 1 | 50 | 71 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100288798
Genomic Map