Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007184.hg.1 |
ENST00000554235.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007184.hg.1 |
ENST00000554235.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007184.hg.1 |
ENST00000554235.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007184.hg.1 |
NR_110062.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007184.hg.1 |
NR_110062.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007184.hg.1 |
NR_110062.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400149843.hg.1 |
ENST00000554235.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400149843.hg.1 |
NR_110062.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400149843.hg.1 |
ENST00000554235.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400149843.hg.1 |
NR_110062.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400149843.hg.1 |
TC1400007184.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1400007184.hg.1 |
ENST00000554235.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007184.hg.1 |
ENST00000554235.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007184.hg.1 |
ENST00000554235.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007184.hg.1 |
NR_110062.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007184.hg.1 |
NR_110062.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007184.hg.1 |
NR_110062.1 |
Refseq |