Assay Details
| Gene Symbol: PAIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006451.4 | NP_006442.2 | 11 | 2519 | 150 | |
| NM_182789.3 | NP_877590.1 | 11 | 2282 | 150 | ||
| NM_183323.2 | NP_899152.1 | 11 | 2087 | 150 | ||
| XM_005248230.3 | XP_005248287.1 | 11 | 2241 | 150 | ||
| XM_017008956.1 | XP_016864445.1 | 11 | 2422 | 150 | ||
| GenBank mRNA | AL136920.1 | - | 11 | 2468 | 150 | |
| DQ786248.1 | - | 1 | 775 | 150 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

PAIP1
Genomic Map