Assay Details
| Gene Symbol: CCSER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207491.2 | NP_997374.1 | 8 | 3355 | 91 | |
| GenBank mRNA | BC051694.1 | - | 8 | 3355 | 5296943 | 91 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

CCSER1
Genomic Map