Assay Details
| Gene Symbol: LINC00315 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF427488.1 | - | 3 | 2252 | 74 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC642852 | LOC642852 | -2.9kb | Sense | Chr. 21 - 45288052 - 45297354 on NCBI Build GRCh38 | |
| Downstream | long intergenic non-protein coding RNA 316 | LINC00316 |
C21orf111, NCRNA00316, PRED59 |
38.1kb | Anti-sense | Chr. 21 - 45338590 - 45341990 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11416207 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11416189 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32079033 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11416204 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11416241 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11416208 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32079097 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32079050 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11416229 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11416202 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32079091 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11416177 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11416246 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11416181 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11416210 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11416244 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11416190 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11416215 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11416230 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17243438 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32079061 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11416231 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11416173 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11416179 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11416193 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32079074 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17243446 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32079047 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11416235 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11416232 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11416237 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11416196 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11416174 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11416225 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11416224 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17243445 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11416220 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11416233 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32079101 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32079046 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32079056 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11416191 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11416176 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11416228 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11416194 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11416214 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32079079 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11416227 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32079022 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11416216 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11416206 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11416185 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11416240 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11416182 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11416184 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11416219 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32079106 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11416172 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 41369 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 18558 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P100190 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 170890 | BC006356.2 | genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 170890 | BC006396.1 | genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 170890 | BC018611.1 | genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 170890 | NM_016170.3 | refseq_mrna_human_NM |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 94047 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03596530 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P131234 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03596537 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03596534 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 8070899 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8070898 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8070897 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8070896 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1553832_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008410.hg.1 | ENST00000441947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008410.hg.1 | ENST00000441947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008410.hg.1 | ENST00000441947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008410.hg.1 | ENST00000441947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008410.hg.1 | ENST00000441947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008410.hg.1 | ENST00000441947.1 | Ensembl |
| Human Genome U133 Plus 2.0 Array | 1553832_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1553832_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553832_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553832_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553832_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553832_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553832_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553832_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553832_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553832_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553832_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553832_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553832_at3 | - | - |
| Human Genome U95 Set | 85453_at | - | - |
| Human Genome U95 Set | 85453_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1752312 | AF427488.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1752312 | AF427488.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1752312 | AF427488.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1752312 | AF427488.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1752312 | AF427488.1 | human_transcriptome |

LINC00315
Genomic Map